Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAAH1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5104R

Description
Transcriptional regulator which functions as a general RNA polymerase II transcription inhibitor. In cooperation with 7SK snRNA sequesters P-TEFb in a large inactive 7SK snRNP complex preventing RNA polymerase II phosphorylation and subsequent transcriptional elongation.Specifications
FAAH1 | |
Polyclonal | |
Unconjugated | |
HEXIM2 | |
4933402L21Rik; hexamethylene bisacetamide inducible 2; hexamethylene bis-acetamide inducible 2; Hexamethylene bis-acetamide-inducible protein 2; hexamethylene-bis-acetamide-inducible transcript 2; hexamthylene bis-acetamide inducible 2; HEXIM2; L3; MAQ1 paralog; Protein HEXIM2; RGD1311635 | |
Rabbit | |
Protein A | |
RUO | |
124790 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96MH2 | |
HEXIM2 | |
KLH conjugated synthetic peptide derived from human FAAH1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction