Learn More
Aviva Systems Biology Corp FAAH2 Antibody - C-terminal region (ARP53305_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53305_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAAH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FAAH2. Alternate names: AMDD, FAAH-2, FLJ31204, RP11-479E16ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60993