Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Fc F(ab ft.)2-Goat anti-Human, PE, Invitrogen™ Non-distribution product as customer accommodation.

F(ab ft.)2-Goat Polyclonal Secondary Antibody

Manufacturer:  Invitrogen H15104

Catalog No. 50-113-7424


Add to cart

Description

Description

Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Human IgM Fc, Fab′2-Goat anti-Human IgM Fc Secondary Antibody, PE
Polyclonal
PBS with BSA, sucrose and 0.1% sodium azide
F(ab′)2-Goat
Purified
ASR
4° C, store in dark
Secondary
Human
Flow Cytometry
PE
Liquid
IgG
0.5 mL
Immunology
Secondary Antibody
Polyclonal
Documents
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit