Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP1/L-FABP Mouse anti-Human, Janelia Fluor 549, Clone: 2G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP104290JF549

Catalog No. NBP14290549

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

FABP1/L-FABPMonoclonalantibody specifically detects FABP1/L-FABP in Human samples.
Specifications

Specifications

FABP1/L-FABP
2G4
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
FABP1
Recombinant human FABP1 (1-127aa) purified from E. coli
Protein G purified
Breast Cancer, Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism
Primary
2168
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Anti-human FABP1 mAb, clone 2G4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human FABP1 protein.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit