Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FABP1 (L-FABP) Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human FABP1 (Ser 2-Ile 127) fused to polyhistide tag at N-terminus

Manufacturer:  Sino Biological™ 12353H07E5

 View more versions of this product

Catalog No. 12-353-H07E5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human FABP1 (Ser 2-Ile 127) fused to polyhistide tag at N-terminus

  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH: 8.3, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Description & Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
2168
≥90% by SDS-PAGE
Human
Fatty Acid Binding Protein 1, liver
Assay Standard
E.coli
His Tag
Lyophilized
15.6kd
5μg
Full Length, Recombinant
FABP1
Other Proteins
FABP1 (L-FABP) Recombinant Human Protein
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit