Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121247

This item is not returnable.
View return policy
Description
Positive test controls include: Jurkat, Rat spinal cord, Rat testis, Rat eye.
FABP12 Polyclonal antibody specifically detects FABP12 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
FABP12 | |
Polyclonal | |
Unconjugated | |
FABP12 | |
1700008G05Rik; Fabp12; FABP12 {ECO:0000303; fatty acid binding protein 12; fatty acid binding protein ORF; fatty acid-binding protein 12; fatty acid-binding protein 12 {ECO:0000303; intracellular fatty acid-binding protein FABP12; Probable fatty acid-binding protein ENSP00000353650; probable fatty acid-binding protein ENSP00000353650 homolog; PubMed:18786628}; RGD1565000 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human FABP12 (NP_001098751.1). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
3.76 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A6NFH5, B7SUM8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
499570, 646486 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title