Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP2/I-FABP Mouse anti-Human, Clone: CPTC-FABP2-3, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen FABP2/I-FABP
Clone CPTC-FABP2-3
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121884
View Documents
Novus Biologicals
NBP30720520UG
20 μg
Each for $138.37
Add to cart
 
NB121883
View Documents
Novus Biologicals
NBP30720510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

FABP2/I-FABP
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Recombinant full length human FABP2/I-FABP protein
Monoclonal
Protein A or G purified
CPTC-FABP2-3
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cholesterol Metabolism, Lipid and Metabolism
10 mM PBS with 0.05% BSA
2169
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit