Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP2/I-FABP Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2C4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270699JF549

Catalog No. N270699J549


Add to cart

Description

Description

FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

FABP2/I-FABP
OTI2C4
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
FABP2
Full length recombinant protein of human FABP2/I-FABP (NP_000125) produced in HEK293T cells.
Immunogen affinity purified
Cholesterol Metabolism, Lipid and Metabolism
Primary
2169.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit