Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP2 Mouse anti-Human, Clone: OTI2C4, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503574
Description
The intracellular fatty acid-binding proteins belong to a multigene family with nearly twenty identified members. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15 kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Intestinal fatty acid-binding protein 2 gene contains four exons and is an abundant cytosolic protein in small intestine epithelial cells. This gene has a polymorphism at codon 54 that identified an alanine-encoding allele and a threonine-encoding allele. Thr-54 protein is associated with increased fat oxidation and insulin resistance.Specifications
FABP2 | |
Monoclonal | |
0.34 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P12104 | |
FABPI, I-FABP | |
Mouse | |
IgG2b | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
2169 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2C4 | |
Unconjugated | |
FABP2 | |
Liquid | |
FABP2 | |
Full length human recombit protein of human FABP2 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title