Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP3/H-FABP Mouse anti-Human, Clone: 9F10, Novus Biologicals™
Mouse Monoclonal Antibody
$337.00 - $482.50
Specifications
Antigen | FABP3/H-FABP |
---|---|
Clone | 9F10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, ELISA 1:100 - 1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6023450UL
|
Novus Biologicals
NBP26023450UL |
50 μL |
Each for $337.00
|
|
|||||
NB60234100U
|
Novus Biologicals
NBP260234100UL |
100 μL |
Each for $482.50
|
|
|||||
Description
FABP3/H-FABP Monoclonal specifically detects FABP3/H-FABP in Human samples. It is validated for Western Blot, ELISA.Specifications
FABP3/H-FABP | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
2170 | |
Recombinant human FABP3 (1-133aa) purified from E. coli | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
9F10 | |
Western Blot 1:1000, ELISA 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
FABP3 | |
IgG1 κ | |
Protein A purified |
RUO
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title