Sino Biological™ FABP3 Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human FABP3 (Met 1-Ala 133)

Manufacturer: Sino Biological™ 12476HNAE250

 View more versions of this product

Catalog No. 12-476-HNAE25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 14.9kd
    Quantity 5 x 50μg
    Common Name Fatty Acid Binding Protein 3, muscle and heart (mammary-derived growth inhibitor)
    Gene Symbol FABP3
    Protein Tag Untagged
    Product Type FABP3 Recombinant Human Protein
    For Use With (Application) Analysis of protein stucture and used in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 2170
    Purity ≥90% by SDS-PAGE
    Source Human
    Structural Form Full Length, Recombinant
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System E.coli

    Expressed from DNA sequence encoding human FABP3 (Met 1-Ala 133)

    • Formulation: 50mM Tris, pH 8.2, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit