Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP4 Polyclonal Antibody, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA121216

Catalog No. PIPA121216


Add to Cart

Description

Description

PA1-21216 detects Fatty-Acid Binding Protein in Human, Mouse and Rat samples.

FABP4 Polyclonal antibody specifically detects FABP4 in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FABP4
Polyclonal
Unconjugated
FABP4
3T3-L1 lipid-binding protein; 422/aP2; adipocyte fatty acid binding protein; Adipocyte lipid binding protein; adipocyte lipid-binding protein; adipocyte protein aP2; adipocyte-type fatty acid-binding protein; AFABP; A-FABP; ALBP; ALBP/Ap2; Ap2; epididymis secretory protein Li 104; FABP4; fatty acid binding protein 4; Fatty acid binding protein 4 adipocyte; fatty acid binding protein 4, adipocyte; fatty acid-binding protein 4; Fatty acid-binding protein, adipocyte; HEL S 104; HEL-S-104; Lbpl; myelin P2 protein homolog; P15; P2 adipocyte protein; Protein 422
Chicken
Protein L
RUO
11770, 2167, 79451
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P04117, P15090, P70623
FABP4
Synthetic peptides: FDEVTADDRKVK and KWDGKSTTIKRK, corresponding to amino acids 70-81 and 96-107 of ALBP
500 μg
Primary
Human, Mouse, Rat
Antibody
IgY
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit