Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FABP4 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding mouse FABP4 (Met 1-Ala 132) fused to polyhistidine tag at C-terminus

Manufacturer:  Sino Biological™ 50652M08H5

 View more versions of this product

Catalog No. 50-652-M08H5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding mouse FABP4 (Met 1-Ala 132) fused to polyhistidine tag at C-terminus

  • Formulation: 50mM Tris, 20% glycerol, pH 9.5
Description & Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
11770
>85% by SDS-PAGE
Mouse
<1 EU/μg
FABP4
Other Proteins
FABP4 Recombinant Mouse Protein
Lyophilized
16kd
5μg
Full Length, Recombinant
Fatty Acid Binding Protein 4 adipocyte
Assay Standard
Mammalian
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit