Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP5/E-FABP, Mouse anti-Human, Clone: FABP5/3750, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | FABP5/E-FABP |
---|---|
Clone | FABP5/3750 |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry 0.1-0.2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
Description
FABP5/E-FABP Monoclonal specifically detects FABP5/E-FABP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
FABP5/E-FABP | |
0.2 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
EFABP, E-FABPPAFABP, fatty acid binding protein 5 (psoriasis-associated), Fatty acid-binding protein 5, PA-FABPepidermal, Psoriasis-associated fatty acid-binding protein homolog | |
FABP5 | |
IgG2a κ | |
Protein A or G purified |
FABP5/3750 | |
Immunohistochemistry 0.1-0.2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q01469, Q01469 | |
2171 | |
Recombinant full-length human FABP5/E-FABP protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title