Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FABP9/T-FABP Mouse anti-Human, Alexa Fluor™ 532, Clone: 13F9, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP231200AF532
Description
FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.Specifications
FABP9/T-FABP | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
FABP9 | |
Recombinant human FABP9 (1-132aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA | |
13F9 | |
Western Blot, ELISA | |
FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP | |
Mouse | |
Protein A purified | |
Developmental Biology, metabolism | |
646480.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title