Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FABP9/T-FABP Mouse anti-Human, Alexa Fluor™ 750, Clone: 13F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP231200AF750

Catalog No. NP23120A750


Add to cart

Description

Description

FABP9/T-FABP Monoclonal antibody specifically detects FABP9/T-FABP in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

FABP9/T-FABP
13F9
Western Blot, ELISA
Conjugated, Purified
FABP9
Recombinant human FABP9 (1-132aa) purified from E. coli
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, T-FABP, TLBP
Mouse
IgG1 Kappa
0.1 mL
Primary
646480.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit