Learn More
Fisherbrand™ Fabric Chair Chrome - Desk Height with Seat Tilt, Adjustable Arms, and Casters in Fabric Products for Science Education
Fisherbrand™ Fabric Task Chairs in Chrome are height adjustable for 31 in. work surface and feature ergonomic adjustments, adjustable arms,and self-braking casters. Heavy-duty olefin fabric in a variety of colors will suit any work space.
$399.00
Specifications
Adjustable Yes (Pneumatic)
Footrest No
With Armrest Yes
With Backrest Yes
With Casters Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36285 Products for Science Education
View Documents
Blue
Each for $399.00
Add to cart
S36523 Products for Science Education
View Documents
Black
Each for $399.00
Add to cart
S36524 Products for Science Education
View Documents
Burgundy
Each for $399.00
Add to cart
S36433 Products for Science Education
View Documents
Gray
Each for $399.00
Add to cart
S37028 Products for Science Education
View Documents
Navy
Each for $399.00
Add to cart
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic seat height adjustment
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Heavy-duty fabric.
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Yes
Yes