Learn More
Fisherbrand™ Fabric Chair Chrome - High Bench Height with Seat Tilt, Chrome Foot Ring, and Casters Products for Science Education
Fisherbrand™ Fabric Task Chairs in Chrome are height adjustable for 31 in. work surface and feature ergonomic adjustments, chrome adjustable foot ring, and self-braking casters. Heavy-duty olefin fabric in a variety of colors will suit any work space.
$398.00
Specifications
Adjustable Yes (Pneumatic)
Footrest Yes
With Armrest No
With Backrest Yes
With Casters Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36001 Products for Science Education
View Documents
Blue
Each for $398.00
Add to cart
S38548 Products for Science Education
View Documents
Black
Each for $398.00
Add to cart
S36974 Products for Science Education
View Documents
Burgundy
Each for $398.00
Add to cart
S36003 Products for Science Education
View Documents
Gray
Each for $398.00
Add to cart
S38435 Products for Science Education
View Documents
Navy
Each for $398.00
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic seat height adjustment
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Heavy-duty fabric
Specifications
Specifications
Yes (Pneumatic)
No
Yes
Yes
Yes
Yes