Learn More
Fisherbrand™ Fabric Chair Chrome - High Bench Height with Seat Tilt, Chrome Foot Ring, and Stationary Glides in Fabric
SCI_ED
Fisherbrand™ Fabric Task Chairs in Chrome are height adjustable for 31 in. work surface and feature ergonomic adjustments, chrome adjustable foot ring, and stationary glides. Heavy-duty olefin fabric in a variety of colors will suit any work space.
 View more versions of this product
Catalog No. S38546
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic seat height adjustment
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Heavy-duty fabric.
Specifications
Specifications
Standard
Navy
Black-non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
High Bench Height
15 year parts, lifetime on pneumatic lift, 5 year on upholstery
19 in.
No
No
26.25 to 36 in.
General Seating Chair
Yes (Pneumatic)
18 in.
Yes
Round tube chrome foot ring with internal lock
Polyurethane foam, Olefin
2.5 in.
37 lb.
25 in.
Yes
Yes
Polished Cast Aluminum