Learn More
Fisherbrand™ Fabric Chair - High Bench Height with Medium Back, Seat Tilt, Adjustable Arms, Chrome Foot Ring, and Stationary Glides in Fabric Products for Science Education
Fisherbrand™ Fabric Task Chairs with Medium Back are height adjustable for 42 in. work surface and feature ergonomic adjustments, larger back, arms, chrome adjustable foot ring, and stationary glides. Heavy-duty olefin fabric in a variety of colors will suit any work space.
$422.00
Specifications
Adjustable Yes (Pneumatic)
Footrest Yes
With Armrest Yes
With Backrest Yes
With Casters No
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36130 Products for Science Education
View Documents
Blue
Each for $422.00
Add to cart
S38469 Products for Science Education
View Documents
Black
Each for $422.00
Add to cart
S37008 Products for Science Education
View Documents
Burgundy
Each for $422.00
Add to cart
S38468 Products for Science Education
View Documents
Gray
Each for $422.00
Add to cart
S37009 Products for Science Education
View Documents
Navy
Each for $422.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 25-34.5 inches from the top of the seat to the floor
  • Features ergonomic seat height adjustment
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Black, nylon-reinforced fiberglass base
  • Stationary, non-marring glides suitable for hard floors
  • Heavy-duty fabric
Specifications
Specifications
Yes (Pneumatic)
Yes
No
Yes
Yes
Yes