Learn More
Fisherbrand™ Fabric Chair - Medium Bench Height with Medium Back, Seat Tilt, Adjustable Arms, Chrome Foot Ring, and Casters in Fabric
Fisherbrand™ Fabric Task Chairs with Medium Back are height adjustable for 36 in. work surface and feature ergonomic adjustments, larger back, arms, chrome adjustable foot ring, and self-braking casters. Heavy-duty olefin fabric in a variety of colors will suit any work space.
$569.00
Specifications
Adjustable Yes (Pneumatic)
Base Material Nylon Reinforced Fiberglass
Seat Material Polyurethane foam, Olefin
Footrest Yes
With Armrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-360-736
View Documents
Blue
Each for $569.00
Add to cart
14-360-738
View Documents
Black
Each for $569.00
Add to cart
14-360-739
View Documents
Burgundy
Each for $569.00
Add to cart
14-360-737
View Documents
Gray
Each for $569.00
Add to cart
14-360-740
View Documents
Navy
Each for $569.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21-28.25 inches from the top of the seat to the floor
  • Features ergonomic seat height adjustment
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Heavy-duty fabric
Specifications
Specifications
Yes (Pneumatic)
Polyurethane foam, Olefin
Yes
Yes
Nylon Reinforced Fiberglass
Yes
Yes
Medium Bench Height