Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Fabric ESD Chair - Desk Height with ESD Stationary Glides in ESD Fabric
Fisherbrand™ Fabric ESD Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments and electro-static components such as fabric and stationary glides. Chairs are manufactured to ensure proper grounding for static sensitive applications.
Specifications
Adjustable Yes
Footrest No
With Armrest No
With Casters No
With Tilt No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-400-209
View Documents
Blue
Each for $472.00
N/A
14-400-208
View Documents
Black
Each for $472.00
N/A
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Yes
No
No
No
No
ANSI/ESD-STM-12.1-2006