Learn More
Fisherbrand™ Fabric ESD Chair - High Bench Height with Adjustable Arms, Chrome Foot Ring, and ESD Casters in ESD Fabric
Fisherbrand™ Fabric ESD Chairs are height adjustable for 42 in. work surfaces & feature ergonomic adjustments, electro-static components such as fabric & self-braking casters, & adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$679.50
Specifications
Adjustable Yes
Footrest Yes
With Armrest Yes
With Backrest Yes
With Casters Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-400-253
View Documents
Blue
Each for $679.50
Add to cart
14-400-252
View Documents
Black
Each for $679.50
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Yes
Yes
Yes
ANSI/ESD-STM-12.1-2006
Yes
Yes
No