Learn More
Fisherbrand™ Fabric ESD Chair - High Bench Height with Medium Back, Chrome Foot Ring, and ESD Casters in ESD Fabric
Fisherbrand™ Fabric ESD Chairs with Medium Back are height adjustable for 42 in. work surfaces & feature ergonomic adjustments & electro-static components such as fabric & self-braking casters. Chairs with Medium Back are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. 14-000-470
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Chrome adjustable foot ring
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Standard
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
20 in.
Polyurethane foam, heavy-duty ESD fabric
37 lb.
Yes
No
300 lb.
ESD Chair
Yes
Black
Yes
Round tube chrome foot ring with internal lock
High Bench Height
No
Yes
26.25 to 36 in.
Polished Cast Aluminum
136 kg