Learn More
Fisherbrand™ Fabric ESD Chair - High Bench Height with Medium Back, Seat Tilt, Adjustable Arms, Chrome Foot Ring, and ESD Casters in ESD Fabric
SCI_ED
Fisherbrand™ Fabric ESD Chairs with Medium Back are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, electro-static components such as fabric and self-braking casters, and arms. Chairs with Medium Back are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. S40340
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Standard
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Black
Yes
Round tube chrome foot ring with internal lock
Polyurethane foam, heavy-duty ESD fabric
48 lb.
Yes
Yes
300 lb.
ESD Chair
Yes
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
20 in.
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
High Bench Height
Yes
Yes
26.25 to 36 in.
Polished Cast Aluminum
136 kg