Learn More
Fisherbrand™ Fabric ESD Chair - High Bench Height with Seat Tilt, Chrome Foot Ring, and ESD Stationary Glides in ESD Fabric Products for Science Education
Fisherbrand™ Fabric ESD Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments and electro-static components such as fabric and stationary glides. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$500.00
Specifications
Adjustable Yes
Footrest Yes
With Armrest No
With Backrest Yes
With Tilt Yes
View More Specs
Products 2
Catalog Number Color Foot Type Price Quantity & Availability  
Catalog Number Color Foot Type Price Quantity & Availability  
S36201 Products for Science Education
View Documents
Black Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Each for $500.00
Add to cart
S36379 Products for Science Education
View Documents
Blue Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Each for $500.00
Add to cart
Description
Description
  • Adjustable from 26.25-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Yes
No
Yes
Yes
Yes
ANSI/ESD-STM-12.1-2006