Learn More
Fisherbrand™ Fabric ESD Chair - Medium Bench Height with Adjustable Arms and ESD Casters in ESD Fabric
SCI_ED
Fisherbrand™ Fabric ESD Chairs are height adjustable for 36 in. work surfaces & feature ergonomic adjustments, electro-static components such as fabric & self-braking casters, & adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. S38649
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Electro-static dissipating components to ensure proper grounding
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Standard
6-way adjustable (move up and down, in and out, and twist toward or away from the user)
Blue
No
Medium Bench Height
Yes
Yes
21.5 to 29 in.
ESD Chair
Yes
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
Polyurethane foam, heavy-duty ESD fabric
42 lb.
Yes
No
Polished Cast Aluminum