Learn More
Fisherbrand Fabric ESD Chair - Medium Bench Height with Chrome Foot Ring and ESD Stationary Glides in ESD Fabric
Fisherbrand Fabric ESD Chairs are height adjustable for 36 in. work surfaces and feature ergonomic adjustments and electro-static components such as fabric and stationary glides. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$439.00
Specifications
Adjustable | Yes |
---|---|
Base Style | Polished Cast Aluminum |
Material | Polyurethane foam cushioning covered with static-dissipating, heavy duty fabric |
Footrest | Yes |
With Armrest | No |
Description
- Adjustable from 21.5-29 inches from the top of the seat to the floor
- Features ergonomic adjustments including seat height
- Locking back tilt
- 20-Inch Chrome adjustable foot ring with internal lock
- Electro-static dissipating components to ensure proper grounding
- Cast-aluminum base
- Stationary ESD glides suitable for hard floors
- Electro-static dissipating fabric
- Meets ANSI/ESD STM12.1-2013
Specifications
Yes | |
Polyurethane foam cushioning covered with static-dissipating, heavy duty fabric | |
No | |
No | |
Medium Bench Height |
Polished Cast Aluminum | |
Yes | |
Yes | |
No | |
ANSI/ESD-STM-12.1-2006 |