Learn More
Fisherbrand™ Fabric ESD Chair - Medium Bench Height with Medium Back and ESD Casters in ESD Fabric
Fisherbrand™ Fabric ESD Chairs with Medium Back are height for 36 in. work surfaces and feature ergonomic adjustments, electro-static components such as fabric and self-braking casters, and arms. Chairs with Medium Back are manufactured to ensure proper grounding for static sensitive applications.
$502.00
Specifications
Adjustable Yes
Footrest No
With Armrest No
With Backrest Yes
With Casters Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-432
View Documents
Blue
Each for $502.00
Add to cart
14-000-438
View Documents
Black
Each for $502.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Yes
No
Yes
ANSI/ESD-STM-12.1-2006
No
Yes
No