Learn More
Fisherbrand™ Fabric ESD Chair - Medium Bench Height with Medium Back, Seat Tilt, and ESD Stationary Glides in ESD Fabric
SCI_ED
Fisherbrand™ Fabric ESD Chairs with Medium Back are height for 36 in. work surfaces and feature ergonomic adjustments, electro-static components such as fabric and stationary glides, and arms. Chairs with Medium Back are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. S40311
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating fabric
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Standard
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
Medium Bench Height
No
No
21.5 to 29 in.
Polished Cast Aluminum
136 kg
Yes
Black
No
Polyurethane foam, heavy-duty ESD fabric
34 lb.
Yes
Yes
300 lb.
ESD Chair