Learn More
MSA™ V-Gard™ Foldback Face Shield Frames
Securely hold face shields, visors, and welding shields
Supplier: MSA™ 10021614
Description
- Durable plastic frame for good dielectric ability
- Easily attaches to helmets
- Snap-on visor attachment secures visor to helmet
Order Info
Order visors separately.
Compliance
Meet ANSI™ Z89.1-2005 standards.