Learn More
CederlaneSupplier Diversity Partner FACTOR 11C,(REACTS WITH ANTIGE  DFS Item
Manufacturer:  CederlaneSupplier Diversity Partner 16944(SS)
Factor 11c,(Reacts with antigen:11A, 11C, 11D),(No. of tests: 300),(SSI® PNEUMOCOCCUS FACTOR ANTISERA FOR THE CAPSULAR REACTION TEST (NEUFELD))
Catalog No. NC1480571
Please call customer service for current pricing. 1-800-766-7000