Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor IX Mouse anti-Human, Alexa Fluor™ 750, Clone: 1C2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP223553AF750

Catalog No. NP2233A70


Add to cart

Description

Description

Factor IX Monoclonal antibody specifically detects Factor IX in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

Factor IX
1C2
Western Blot, ELISA
Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Clone 1C2 is specific for native human factor IX
ELISA, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
F9
Factor IX isolated from human plasma adsorbed onto aluminum hydroxide gel
0.1 mL
Primary
2158
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit