Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor IX Mouse anti-Human, Clone: 13B9, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Factor IX
Clone 13B9
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:4000
Applications Western Blot, ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP105079
View Documents
Novus Biologicals
NBP105079
100 μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Factor IX Monoclonal antibody specifically detects Factor IX in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Factor IX
1.0 mg/mL
Western Blot, ELISA
Unconjugated
Mouse
Human
2158
Factor IX isolated from human plasma adsorbed onto aluminum hydroxide gel
Primary
Store at 4C in the dark.
13B9
Western Blot 1:100-1:2000, ELISA 1:4000
Monoclonal
Purified
RUO
Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
F9
IgG1 κ
Protein A or G purified
Factor IX (human, Christmas factor)
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit