Learn More
Factor IX Rabbit anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP299803AF700
Catalog No. NB062993
Add to cart
Description
Description
Factor IX Polyclonal antibody specifically detects Factor IX in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
Factor IX
Alexa Fluor 700
50mM Sodium Borate
Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
Produced in rabbits immunized with purified, recombinant Factor IX (AAB59620.1; Met 1-Thr 461).
Affinity purified
RUO
Primary
2158
ELISA, Western Blot
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human