Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor VIII Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP299651AF488
Description
Factor VIII Polyclonal antibody specifically detects Factor VIII in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Factor VIII | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Factor VIII. | |
Antigen Affinity-purified | |
RUO | |
Primary | |
2157 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Store at 4°C in the dark. | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title