Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Rabbit anti-Human, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299651IR

Catalog No. NB062303


Add to cart

Description

Description

Factor VIII Polyclonal antibody specifically detects Factor VIII in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Factor VIII
DyLight 755
50mM Sodium Borate
AHF, Antihemophilic factor, coagulation factor VIII, procoagulant component, coagulation factor VIIIc, DXS1253E, F8Ccoagulation factor VIII, factor VIII F8B, FVIII, HEMAF8B, Procoagulant component
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Factor VIII.
Antigen Affinity-purified
RUO
Primary
2157
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit