Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Factor VIII Related Antigen/von Willebrand Factor Ab-1, Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

$189.00 - $410.50

Specifications

Antigen Factor VIII Related Antigen/von Willebrand Factor Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RB281A
View Documents
Thermo Scientific™
RB281A
1mL Each for $410.50
Add to cart
 
RB281A0
View Documents
Thermo Scientific™
RB281A0
100μL Each for $189.00
Add to cart
 
RB281A1
View Documents
Thermo Scientific™
RB281A1
500μL Each for $215.50
Add to cart
 
RB281R1
View Documents
Thermo Scientific™
RB281R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
RB281R7
View Documents
Thermo Scientific™
RB281R7
7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Factor VIII Related Antigen/von Willebrand Factor Ab-1, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Factor VIII related antigen or von Willebrand factor is a multimeric glycoprotein. It has functional binding domains to platelet glycoprotein Ib, glycoprotein IIb/IIIa, collagen and heparin. von Willebrand factor is synthesized by endothelial cells and stored in the Weibel-Palade granules. It mediates platelet adhesion to injured vessel walls and serves as a carrier and stabilizer for coagulation factor VIII. von Willebrand factor is one of the most useful markers to identify endothelial (or megakaryocytic) lineage of neoplasms. As not all endothelial cells synthesize/store this molecule, about 30% of tumors of vascular origin fail to stain for factor VIII related antigen, regardless of whether they are benign or malignant. Staining for factor VIII related antigen has also been used to measure angiogenesis an indicator of tumor recurrence.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified factor VIII related antigen/von Willebrand factor

Molecular Weight: 270 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Specifications

Specifications

Factor VIII Related Antigen/von Willebrand Factor Ab-1
Polyclonal
Rabbit
Hematopoietic
Purified factor VIII related antigen/von Willebrand factor
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit