Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor VIII Related Antigen/von Willebrand Factor Ab-1 Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

Supplier:  Epredia RB281A0

 View more versions of this product

Catalog No. RB281A0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

Factor VIII related antigen or von Willebrand factor is a multimeric glycoprotein. It has functional binding domains to platelet glycoprotein Ib, glycoprotein IIb/IIIa, collagen and heparin. von Willebrand factor is synthesized by endothelial cells and stored in the Weibel-Palade granules. It mediates platelet adhesion to injured vessel walls and serves as a carrier and stabilizer for coagulation factor VIII. von Willebrand factor is one of the most useful markers to identify endothelial (or megakaryocytic) lineage of neoplasms. As not all endothelial cells synthesize/store this molecule, about 30% of tumors of vascular origin fail to stain for factor VIII related antigen, regardless of whether they are benign or malignant. Staining for factor VIII related antigen has also been used to measure angiogenesis an indicator of tumor recurrence.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified factor VIII related antigen/von Willebrand factor

Molecular Weight: 270 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
Specifications

Specifications

Factor VIII Related Antigen/von Willebrand Factor Ab-1
Polyclonal
Rabbit
100 μL
Hematopoietic
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Purified factor VIII related antigen/von Willebrand factor
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit