Learn More
Factor VIII Related Antigen/von Willebrand Factor Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments.
$485.00 - $643.00
Specifications
Antigen | Factor VIII Related Antigen/von Willebrand Factor Ab-2 |
---|---|
Clone | F8/86 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS722P1ABX
|
Epredia
MS722P1ABX |
Concentrated | 100 μL |
Each for $543.00
|
|
||||
MS722PABX
|
Epredia
MS722PABX |
Concentrated | 200 μL |
Each for $643.00
|
|
||||
MS722P1
|
Epredia
MS722P1 |
Concentrated | 500 μL |
Each for $485.00
|
|
||||
MS722P
|
Epredia
MS722P |
Concentrated | 1 mL |
Each for $591.00
|
|
||||
MS722P0
|
Epredia
MS722P0 |
Concentrated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS722R1
|
Epredia
MS722R1 |
Prediluted | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS722RQ
|
Epredia
MS722RQ |
Ready-to-Use | 12 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
Factor VIII related antigen or von Willebrand factor is a multimeric glycoprotein, which binds to platelet glycoprotein Ib, glycoprotein IIb/IIIa, collagen and heparin. It is synthesized by endothelial cells and stored in the Weibel-Palade granules. It mediates platelet adhesion to injured vessel walls and serves as a carrier and stabilizer for coagulation factor VIII. von Willebrand factor is one of the most useful markers to identify endothelial (or megakaryocytic) lineage of neoplasms. As not all endothelial cells synthesize/store this molecule, about 30% of tumors of vascular origin fail to stain for factor VIII related antigen, regardless of whether they are benign or malignant. Staining for factor VIII related antigen has also been used to measure angiogenesis, an indicator of tumor recurrence.Host Species: Mouse
Clone: F8/86
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Partially purified von Willebrand factor from human placenta
Molecular Weight: 270 kDa
Positive Control: Blood vessels in tonsil or intestine
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Factor VIII Related Antigen/von Willebrand Factor Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Partially purified von Willebrand factor from human placenta |
F8/86 | |
Monoclonal | |
Mouse | |
Hematopoietic | |
Primary |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.