Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XII heavy chain Mouse anti-Human, Alexa Fluor™ 750, Clone: 10-11-5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP241375AF750

Catalog No. NP24137A70


Add to cart

Description

Description

Factor XII heavy chain Monoclonal antibody specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Factor XII heavy chain
10-11-5
Western Blot, ELISA, Sandwich ELISA
Purified, Conjugated
F12
Affinity purified human factor XII.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Antibody binds the heavy chain of human factor XII, including factor XII derivatives such as a-factor XIIa and a-factor XIIa C1-inhibitor complexes.
Western Blot, ELISA
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor
Mouse
IgG1 Kappa
100 μL
Primary
2161.0
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit