Learn More
Factor XII heavy chain Mouse anti-Human, DyLight 650, Clone: 13799, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241377C
Catalog No. NBP241377C
Add to cart
Description
Description
Factor XII heavy chain Monoclonal antibody specifically detects Factor XII heavy chain in Human samples. It is validated for Western Blot.
Specifications
Specifications
Factor XII heavy chain
13799
Western Blot
Purified
F12
Affinity purified human factor XII.
0.1 ml
Primary
2161
Human
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
beta-factor XIIa part 1, coagulation factor XII, coagulation factor XII (Hageman factor), coagulation factor XIIa heavy chain, coagulation factor XIIa light chain, EC 3.4.21, EC 3.4.21.38, HAE3, HAEX, HAFbeta-factor XIIa part 2, Hageman factor
Mouse
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Antibody binds the heavy chain of human factor XII, including factor XII derivatives such as a-factor XIIa and a-factor XIIa C1-inhibitor complexes.