Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XIII Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270697AF700

Catalog No. N270697A700


Add to cart

Description

Description

Factor XIII Monoclonal antibody specifically detects Factor XIII in Human samples. It is validated for Western Blot.
Specifications

Specifications

Factor XIII
OTI1A3
Western Blot
Affinity Purified, Conjugated
F13B
Recombinant protein fragment corresponding to amino acids 357-661 of human Factor XIII (NP_001985) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma
Mouse
IgG1
0.1 mL
Primary
2165.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit