Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIa Mouse anti-Human, Clone: F13A1/1447, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP253183100UG
Description
Factor XIIIa Monoclonal antibody specifically detects Factor XIIIa in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page.Specifications
Factor XIIIa | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page | |
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma | |
Mouse | |
83 kDa | |
100 μg | |
Apoptosis, Cancer, Cell Biology | |
2162 | |
Purified |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page | |
F13A1/1447 | |
Unconjugated | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
F13A1 | |
Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
IgG2a |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only