Learn More
Factor XIIIa Mouse anti-Human, Clone: F13A1/1447, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP25318320UG
 View more versions of this product
Catalog No. NB25318320
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Factor XIIIa Monoclonal antibody specifically detects Factor XIIIa in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, SDS-Page.
Specifications
Specifications
Factor XIIIa
Monoclonal
0.2 mg/mL
Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, SDS-Page
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma
Mouse
83 kDa
20 μg
Apoptosis, Cancer, Cell Biology
2162
Purified
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page
F13A1/1447
Unconjugated
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
F13A1
Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
Human
IgG2a