Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Factor XIIIa Mouse anti-Human, Clone: OTI6F5, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270696
Description
Factor XIIIa Monoclonal antibody specifically detects Factor XIIIa in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Factor XIIIa | |
OTI6F5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma | |
Mouse | |
IgG1 | |
Immunogen affinity purified | |
Apoptosis, Cancer, Cell Biology | |
Primary | |
2162 | |
Human |
CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot | |
LYOPH | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready | |
Affinity Purified, Lyophilized | |
F13A1 | |
Full length human recombinant protein of human F13A1 (NP_000120) produced in HEK293T cell. | |
79.2 kDa | |
100 μg | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title