Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Factor XIIIa Mouse anti-Human, PE/Atto594, Clone: SPM180, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254480PEATT594

Catalog No. N254480P594


Add to cart

Description

Description

Factor XIIIa Monoclonal antibody specifically detects Factor XIIIa in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Factor XIIIa
SPM180
Flow Cytometry
Conjugated, Purified
F13A1
Recombinant fragment of human Factor XIIIa protein (aa46-181) (exact sequence is proprietary)
Protein A or G purified
Apoptosis, Cancer, Cell Biology
Primary
2162.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma
Mouse
IgG2b Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit