Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1C11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271387AF647

Catalog No. N271387A647


Add to cart

Description

Description

FADD Monoclonal antibody specifically detects FADD in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

FADD
OTI1C11
Western Blot, Flow Cytometry
Affinity Purified, Conjugated
FADD
Full length recombinant protein of human FADD (NP_003815) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors
Primary
8772.0
Flow Cytometry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit