Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FADD Mouse anti-Human, PerCP, Clone: OTI1C11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271387PCP
Description
FADD Monoclonal specifically detects FADD in Human samples. It is validated for Flow Cytometry.Specifications
FADD | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
FADD | |
Full length recombinant protein of human FADD (NP_003815) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI1C11 | |
Flow Cytometry | |
Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors | |
8772.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title