Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FADD Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen FADD
Applications Immunohistochemistry, Immunoprecipitation, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092519
View Documents
Novus Biologicals
NBP303350100UL
100 μg
Each for $320.71
Add to cart
 
NB092520
View Documents
Novus Biologicals
NBP30335020UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

FADD Polyclonal antibody specifically detects FADD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown
Specifications

Specifications

FADD
Unconjugated
Rabbit
Apoptosis, Cancer, Death Receptor Signaling Pathway, Diabetes Research, Necroptosis, Phospho Specific, Tumor Suppressors
PBS with 50% glycerol, pH7.3.
8772
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunoprecipitation, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
Fas (TNFRSF6)-associated via death domain, FAS-associated death domain protein, FAS-associating death domain-containing protein, GIG3, Growth-inhibiting gene 3 protein, Mediator of receptor induced toxicity, mediator of receptor-induced toxicity, MGC8528, MORT1Fas-associating protein with death domain, protein FADD
Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human FADD (NP_003815.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit